missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid, ACS, 85%, Spectrum™ Chemical

Catalog No. 18612337
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Glass Bottle
Poly Drum
Poly-Coated Glass Bottle
Catalog No. Quantity Packaging
18612337 100 mL Amber Glass Bottle
18612341 500 mL Amber Glass Bottle
18612335 1 L Amber Glass Bottle
18612338 2.5 L Glass Bottle
18612339 2.5 L Poly-Coated Glass Bottle
18612336 20 L Poly Drum
18612340 200 L Drum
7 options

Catalog No. 18612337

Supplier: Spectrum Chemical Mfg Cor P1095100ML

Only null left

Chemical Identifiers

CAS 7664-38-2 , 7732-18-5
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.99
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
IUPAC Name phosphoric acid
SMILES OP(O)(O)=O

Specifications

CAS Max % 1
Heavy Metals 1e-05
Packaging Amber Glass Bottle
Reducing Substances To Pass Test
Quantity 100 mL
Percent Purity 85%
Grade ACS

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.