missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoric Acid, 85%, Electronic Grade, 85.0-87.0%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Reducing Substances | To Pass Test |
| Quantity | 1 gal. |
| Percent Purity | 85 to 87% |
| Grade | Histological |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.