missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid, 85%, ACS Reagent (Orthophosphoric Acid), Reagents

Catalog No. p-200021210
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
20 L
Packaging:
Glass Bottle
HDPE Winpak
Poly Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01336824 1 L Poly Bottle
01336823 2.5 L Poly Bottle
01336822 2.5 L Glass Bottle
01336821 20 L HDPE Winpak
01336820 4 L Poly Bottle
01336819 4 L Glass Bottle
01336818 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 01336824 Supplier Labchem Inc Supplier No. C51019501A
Only null left

Phosphoric Acid, 85%, ACS Reagent (Orthophosphoric Acid), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7664-38-2 , 7732-18-5
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.99
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym Orthophosphoric Acid, o-Phosphoric Acid
IUPAC Name phosphoric acid
SMILES OP(O)(O)=O

Specifications

pH <2
Color Colorless
Boiling Point 260°C
Packaging Poly Bottle
Specific Gravity 1.69
Quantity 1 L
Physical Form Liquid
Grade ACS
Chemical Name or Material Phosphoric Acid, 85%, ACS Reagent (Orthophosphoric Acid)