missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoric Acid, 85%, ACS Reagent (Orthophosphoric Acid), Reagents
Phosphoric Acid, 85%, ACS Reagent (Orthophosphoric Acid), ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C51019504C
Specifications
| <2 | |
| Colorless | |
| 260°C | |
| 1.69 | |
| H3O4P | |
| Orthophosphoric Acid, o-Phosphoric Acid | |
| OP(O)(O)=O | |
| 97.99 |
| Phosphoric Acid, 85%, ACS Reagent (Orthophosphoric Acid) | |
| 7664-38-2,7732-18-5 | |
| Glass Bottle | |
| 4 L | |
| Liquid | |
| NBIIXXVUZAFLBC-UHFFFAOYSA-N | |
| phosphoric acid | |
| ACS |
Chemical Identifiers
| 7664-38-2 | |
| 97.99 | |
| Orthophosphoric Acid, o-Phosphoric Acid | |
| OP(O)(O)=O |
| H3O4P | |
| NBIIXXVUZAFLBC-UHFFFAOYSA-N | |
| phosphoric acid |