missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoric Acid, ACS Reagent Grade, Ricca Chemical
Supplier: Ricca Chemical Company RABP003025D1
Specifications
| Phosphoric Acid | |
| 7664-38-2,7732-18-5 | |
| 85% Acid | |
| max. 0.001% | |
| Pass | |
| H3O4P | |
| orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico | |
| OP(=O)(O)O | |
| 97.994 | |
| CHEBI:26078 | |
| ACS |
| Colorless | |
| 85.0 | |
| 100.0 | |
| Glass Bottle | |
| 2.5 L | |
| Liquid | |
| NBIIXXVUZAFLBC-UHFFFAOYSA-N | |
| phosphoric acid | |
| 1004 | |
| ≥85% |
Chemical Identifiers
| 7664-38-2 | |
| NBIIXXVUZAFLBC-UHFFFAOYSA-N | |
| CHEBI:26078 |
| H3O4P | |
| 1004 | |
| phosphoric acid |
Safety and Handling
ShelfLife : 36 months