missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid, ACS Reagent Grade, Ricca Chemical

Catalog No. p-3252070
Change view
Click to view available options
Quantity:
500 mL
2.5 L
4 L
Packaging:
Amber Glass Bottle
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RABP0030500 500 mL Glass Bottle
RABP003025D 2.5 L Glass Bottle
RABP00304C 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RABP0030500 Supplier Ricca Chemical Company Supplier No. RABP0030500D1
Only null left

Chemical Identifiers

CAS 7664-38-2 , 7732-18-5
Molecular Formula H3O4P
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid

Specifications

Color Colorless
Concentration 85% Acid
CAS Min % 85.0
CAS Max % 100.0
Heavy Metals (as Pb) max. 0.001%
Packaging Glass Bottle
Reducing Substances Pass
Quantity 500 mL
Physical Form Liquid
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
SMILES OP(=O)(O)O
Molecular Weight (g/mol) 97.994
Percent Purity ≥85%
Grade ACS
Chemical Name or Material Phosphoric Acid
Show More Show Less
ShelfLife 36 months