missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoric Acid, 75% (w/w), Reagents
Specifications
Specifications
| pH | <2 |
| Color | Colorless |
| Boiling Point | 260°C |
| Packaging | Glass Bottle |
| Specific Gravity | 1.579 |
| Quantity | 2.5 L |
| Physical Form | Liquid |
| Percent Purity | 75% |
| Grade | Reagent |
| Chemical Name or Material | Phosphoric Acid, 75% (w/w) |