missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid, 75% (w/w), Reagents

Catalog No. 01334161
Change view
Click to view available options
Quantity:
2.5 L
4 L
55 gal.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334161 2.5 L
01334160 4 L
01334159 55 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01334161 Supplier Labchem Inc Supplier No. CP2092002.5D1
Only null left

Phosphoric Acid, 75% (w/w), Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 7664-38-2 , 7732-18-5
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.99
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym Orthophosphoric Acid, o-Phosphoric Acid
IUPAC Name phosphoric acid
SMILES OP(O)(O)=O

Specifications

pH <2
Color Colorless
Boiling Point 260°C
Packaging Glass Bottle
Specific Gravity 1.579
Quantity 2.5 L
Physical Form Liquid
Percent Purity 75%
Grade Reagent
Chemical Name or Material Phosphoric Acid, 75% (w/w)