missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid, 55% v/v, Reagents

Catalog No. p-200021735
Change view
Click to view available options
Quantity:
10 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334546 10 L
01334545 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01334546 Supplier Labchem Inc Supplier No. CP00340010F
Only null left

Phosphoric Acid, 55% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7664-38-2 , 7732-18-5
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.99
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym Orthophosphoric Acid, o-Phosphoric Acid
IUPAC Name phosphoric acid
SMILES OP(O)(O)=O

Specifications

pH <2
Color Colorless
Boiling Point 110°C
Packaging Cubitainer
Specific Gravity 1.3795
Quantity 10 L
Physical Form Liquid
Percent Purity 55%
Grade Reagent
Chemical Name or Material Phosphoric Acid, 55% v/v