missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid, 50% v/v, Reagents

Catalog No. 01334170
Change view
Click to view available options
Quantity:
1 L
4 L
10 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334170 10 L
01334169 1 L
01334168 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01334170 Supplier Labchem Inc Supplier No. CP20890010F
Only null left

Phosphoric Acid, 50% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7664-38-2 , 7732-18-5
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.99
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym Orthophosphoric Acid, o-Phosphoric Acid
IUPAC Name phosphoric acid
SMILES OP(O)(O)=O

Specifications

pH <2
Color Colorless
Boiling Point 108°C
Packaging Cubitainer
Specific Gravity 1.363
Quantity 10 L
Physical Form Liquid
Percent Purity 50%
Grade Reagent
Chemical Name or Material Phosphoric Acid, 50% v/v