missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid, 5% (w/w), Reagents

Catalog No. p-200021560
Change view
Click to view available options
Quantity:
20 L
200 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334167 200 L
01334166 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01334167 Supplier Labchem Inc Supplier No. CP209000200E
Only null left

Phosphoric Acid, 5% (w/w), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7664-38-2
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Orthophosphoric Acid, o-Phosphoric Acid
IUPAC Name water
SMILES O

Specifications

pH <2
Color Colorless
Boiling Point 100°C
Packaging Poly Drum
Specific Gravity 1.0272
Quantity 200 L
Physical Form Liquid
Percent Purity 5%
Grade Reagent
Chemical Name or Material Phosphoric Acid, 5% (w/w)