missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoric Acid, 40% w/w, Reagents
Phosphoric Acid, 40% w/w, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.
Supplier: Labchem Inc CP223700500A
Specifications
| <2 | |
| Colorless | |
| 113°C | |
| 1.2536 | |
| H2O | |
| Orthophosphoric Acid, o-Phosphoric Acid | |
| O | |
| 18.02 | |
| Reagent |
| Phosphoric Acid, 40% w/w | |
| 7732-18-5,7664-38-2 | |
| Poly Bottle | |
| 500 mL | |
| Liquid | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| 40% |
Chemical Identifiers
| 7732-18-5 | |
| 18.02 | |
| Orthophosphoric Acid, o-Phosphoric Acid | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |