missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid, 30% (v/v), Ricca Chemical

Catalog No. 5851532
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5851532 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5851532 Supplier Ricca Chemical Company Supplier No. 5851.532

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7732-18-5 , 7664-38-2
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O

Specifications

Color Colorless
CAS Min % 69.0, 29.0
CAS Max % 71.0, 31.0
Packaging Natural Poly Bottle
Quantity 1 L
Physical Form Liquid
Percent Purity 30% V/V
Grade Laboratory
Concentration or Composition (by Analyte or Components) 30% (v/v) Aqueous Solution
Chemical Name or Material Phosphoric Acid
ShelfLife 24 months