missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoric Acid, 20% (w/w), Reagents
Phosphoric Acid, 20% (w/w), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc CP103000500C
Specifications
| <2 | |
| Colorless | |
| 100°C | |
| 1.1155 | |
| H2O | |
| Orthophosphoric Acid, o-Phosphoric Acid | |
| O | |
| 18.02 | |
| Reagent |
| Phosphoric Acid, 20% (w/w) | |
| 7732-18-5,7664-38-2 | |
| Glass Bottle | |
| 500 mL | |
| Liquid | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| 20% |
Chemical Identifiers
| 7732-18-5 | |
| 18.02 | |
| Orthophosphoric Acid, o-Phosphoric Acid | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |