missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid, 20% (v/v), Ricca Chemical

Catalog No. R58503004A
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R58503004A 4 L Natural Poly Bottle
5850316 500 mL Natural Poly Bottle
5850332 1 L Natural Poly Bottle
R585030020F 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. R58503004A Supplier Ricca Chemical Company Supplier No. R58503004A

Chemical Identifiers

CAS 7732-18-5 , 7664-38-2
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O

Specifications

Color Colorless
CAS Min % 79.0, 19.0
CAS Max % 81.0, 21.0
Packaging Natural Poly Bottle
Quantity 4 L
Physical Form Liquid
Percent Purity 20% V/V
Grade Laboratory
Concentration or Composition (by Analyte or Components) 20% (v/v) Aqueous Solution (1 + 4)
Chemical Name or Material Phosphoric Acid
ShelfLife 24 months