missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid, 10% v/v, Reagents

Catalog No. 01334165
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334165 1 L
01334164 20 L
01334163 4 L
01334162 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01334165 Supplier Labchem Inc Supplier No. CP2091001A
Only null left

Phosphoric Acid, 10% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7664-38-2
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Orthophosphoric Acid, o-Phosphoric Acid
IUPAC Name water
SMILES O

Specifications

pH <2
Color Colorless
Boiling Point 100°C
Packaging Poly Bottle
Specific Gravity 1.0754
Quantity 1 L
Physical Form Liquid
Percent Purity 10%
Grade Reagent
Chemical Name or Material Phosphoric Acid, 10% v/v