missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoric Acid, 10% v/v, Reagents
Specifications
Specifications
| pH | <2 |
| Color | Colorless |
| Boiling Point | 100°C |
| Packaging | Poly Bottle |
| Specific Gravity | 1.0754 |
| Quantity | 4 L |
| Physical Form | Liquid |
| Percent Purity | 10% |
| Grade | Reagent |
| Chemical Name or Material | Phosphoric Acid, 10% v/v |