missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid, 10% (v/v), Ricca Chemical

Catalog No. 585025
Change view
Click to view available options
Quantity:
10 L
Packaging:
Cubitainer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
585025 10 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 585025 Supplier Ricca Chemical Company Supplier No. 58502.5

Chemical Identifiers

CAS 7732-18-5 , 7664-38-2
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O

Specifications

Color Colorless
Concentration 10% (v/v)
CAS Min % 89.0, 9.0
CAS Max % 91.0, 11.0
Packaging Cubitainer
Solvent or Matrix Aqueous Solution
Quantity 10 L
Physical Form Liquid
Percent Purity 10% V/V
Grade Laboratory
Chemical Name or Material Phosphoric Acid
Show More Show Less