missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid, 0.050 Molar, Reagents

Catalog No. 01334128
Change view
Click to view available options
Quantity: 20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01334128 Quantity: 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01334128 Supplier Labchem Inc Supplier No. CP22990020F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Phosphoric Acid, 0.050 Molar, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 7664-38-2
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Orthophosphoric Acid, o-Phosphoric Acid
IUPAC Name water
SMILES O

Specifications

pH <2
Color Colorless
Boiling Point 100°C
Packaging Cubitainer
Specific Gravity 1.0027
Quantity 20 L
Physical Form Liquid
Grade Reagent
Chemical Name or Material Phosphoric Acid, 0.050 Molar