missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Phosphoramidite Diluent

Catalog No. 401445
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
401445 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 401445 Supplier Applied Biosystems™ Supplier No. 401445

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Contains 100 mL Phosphoramidite Diluent (anhydrous acetonitrile, DNA synthesis grade).

TestedOur high-performance reagents begin with raw materials that must conform to our strict specifications. Once the reagents are manufactured and packaged, extensive testing is performed to assure you that all liquids are free of interfering particulate matter; reagent concentrations are within our specifications, and the performance specification meets at least 98% stepwise coupling efficiency.
  • Extensive testing and quality controls assure high quality reagents that meet our high expectations and specifications.
  • Reagents and supplies are use-tested and optimized for the 3900 system for quality nucleic acid synthesis.

Specifications

Packaging Type Shipped at Room Temperature
Quantity 100 mL

For Research Use Only. Not for use in diagnostic procedures.