missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphomolybdic acid hydrate, ≥99.99% trace metals basis, MilliporeSigma™ Supelco™
Phosphomolybdic acid hydrate is a heteropolyacid hydrate. It participates in the synthesis of molybdenum disulfide (MoS2)-reduced graphene oxide (RGO) composites. Phosphomolybdic acid is one of the components of the Goldner's trichrome stain used for tissue staining.
Supplier: Merck Emd Millipore 43140050G
Specifications
| 1.62 g/mL (at 25°C) | |
| Glass Bottle | |
| H3 [P(Mo3O10)4 ] · xH2 O | |
| Molybdophosphoric acid | |
| ≥99.99% (trace metals basis) |
| 51429-74-4 | |
| 50 g | |
| MFCD00149913 | |
| 1825.25 (anhydrous basis) |
Chemical Identifiers
| MFCD00149913 | |
| Molybdophosphoric acid |