missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphomolybdic acid hydrate, ≥99.99% trace metals basis, MilliporeSigma™ Supelco™
Specifications
Specifications
| Density | 1.62 g/mL (at 25°C) |
| CAS | 51429-74-4 |
| Packaging | Glass Bottle |
| Quantity | 10 g |
| Linear Formula | H3 [P(Mo3O10)4 ] · xH2 O |
| Formula Weight | 1825.25 (anhydrous basis) |
| Percent Purity | ≥99.99% (trace metals basis) |