missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphomolybdic acid, 20 wt% solution in ethanol
CAS: 51429-74-4 | H3Mo12O40P | 1825.40 g/mol
$132.98 - $132.98
Specifications
| Chemical Name or Material | Phosphomolybdic acid |
|---|---|
| Color | Green to Yellow |
| Name Note | 20 wt% solution in ethanol |
| Density | 0.9400 g/mL |
| CAS | 64-17-5 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC368621000
|
Thermo Scientific Chemicals
368621000 |
100 mL |
Each for $132.98
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentSpecifications
| Phosphomolybdic acid | |
| 20 wt% solution in ethanol | |
| 64-17-5 | |
| 94% | |
| H3Mo12O40P | |
| Solution | |
| 13, 7428 | |
| FEFSFHGZSNHJEL-UHFFFAOYSA-N | |
| phosphoric acid dodecamolybdenum hexatriacontaoxidandiide | |
| 1825.25 |
| Green to Yellow | |
| 0.9400 g/mL | |
| Plastic Bottle | |
| 5°C | |
| H3[P(Mo3O10)4] | |
| MFCD00149913 | |
| Molybdophosphoric acid | |
| [O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].OP(O)(O)=O | |
| 1825.40 | |
| 19 to 21% |
For Manufacturing and Laboratory Use. Not for beverage use