missing translation for 'onlineSavingsMsg'
Learn More

Phospholipid Mixture for HPLC from Soybean (Glycine max), MilliporeSigma™ Supelco™

Catalog No. 111017125
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017125 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017125 Supplier Merck Emd Millipore Supplier No. P38171VL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 67-66-3
Shipping Condition Dry Ice
Physical Form Liquid
Type Certified Reference Material
Quantity 2 mL
Grade Certified Reference Material