missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoenolpyruvic acid trisodium salt heptahydrate, 98%
Description
When coupled with pyruvate kinase it completes an ATP-regenerating system
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| Quantity | 1 g |
| Formula Weight | 360.09 |
| Percent Purity | 98% |
| Physical Form | Powder |
| Chemical Name or Material | Phosphoenolpyruvic acid trisodium salt heptahydrate |
Safety and Handling
| EINECSNumber | 226-906-8 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only