missing translation for 'onlineSavingsMsg'
Learn More

Phosphoenolpyruvic acid trisodium salt heptahydrate, 98%

Catalog No. AAJ6301303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6301303 1 g
AAJ6301306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6301303 Supplier Thermo Scientific Chemicals Supplier No. J6301303

When coupled with pyruvate kinase it completes an ATP-regenerating system | CAS: 5541-93-5 | C3H2Na3O6P | 233.986 g/mol

When coupled with pyruvate kinase it completes an ATP-regenerating system

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5541-93-5
Molecular Formula C3H2Na3O6P
Molecular Weight (g/mol) 233.986
MDL Number MFCD00150737
InChI Key RJUFBDMHZGRGMA-UHFFFAOYSA-K
Synonym 2-propenoic acid, 2-phosphonooxy-, trisodium salt, trisodium 2-phosphonatooxy prop-2-enoate, unii-78y3q07j1x, phosphoenolpyruvate trisodium salt, 2-phosphoenol pyruvate na3salt, 2-propenoic acid, 2-phosphonooxy-, sodium salt 1:3, trisodium phosphonatoenolpyruvate, trisodium 2-phosphonatoacrylate, trisodium phosphoenolpyruvate, c3h2o6p.3na
PubChem CID 79673
IUPAC Name trisodium;2-phosphonatooxyprop-2-enoate
SMILES C=C(C(=O)[O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+]

Specifications

Color White
Quantity 1 g
Formula Weight 360.09
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Phosphoenolpyruvic acid trisodium salt heptahydrate
EINECSNumber 226-906-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only