missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Phosphoenolpyruvic Acid Tris(cyclohexylammonium) Salt Hydrate 98.0+%, TCI America™
$250.51 - $874.29
Chemical Identifiers
CAS | 35556-70-8 |
---|---|
Molecular Formula | C21H44N3O6P |
Molecular Weight (g/mol) | 465.572 |
MDL Number | MFCD00004258 |
InChI Key | MJKYGUXBFYGLLM-UHFFFAOYSA-N |
Synonym | phosphoenolpyruvic acid tris cyclohexylammonium salt, tris cyclohexylamine ; phosphoenolpyruvic acid, phosphoenolpyruvic acid, tris cyclohexylammonium salt, pep-3cha, phosphoenol pyruvate tri cyclohexylammonium salt, phosphoenolpyruvic acid tricyclohexylammonium salt, phosphoenolpyruvic acid tris cyclohexylamine salt, 2-phosphonooxy-2-propenoic acid tri cyclohexylammonium salt, axi c 1/4 +/-ui feminineea ey>>. 1/4 masculine degrees .ni, phospho enol pyruvic acid tri cyclohexylammonium salt enzymatic |
PubChem CID | 11754241 |
IUPAC Name | cyclohexanamine;2-phosphonooxyprop-2-enoic acid |
SMILES | C=C(C(=O)O)OP(=O)(O)O.C1CCC(CC1)N.C1CCC(CC1)N.C1CCC(CC1)N |
Chemical Identifiers
35556-70-8 | |
465.572 | |
MJKYGUXBFYGLLM-UHFFFAOYSA-N | |
11754241 | |
C=C(C(=O)O)OP(=O)(O)O.C1CCC(CC1)N.C1CCC(CC1)N.C1CCC(CC1)N |
C21H44N3O6P | |
MFCD00004258 | |
phosphoenolpyruvic acid tris cyclohexylammonium salt, tris cyclohexylamine ; phosphoenolpyruvic acid, phosphoenolpyruvic acid, tris cyclohexylammonium salt, pep-3cha, phosphoenol pyruvate tri cyclohexylammonium salt, phosphoenolpyruvic acid tricyclohexylammonium salt, phosphoenolpyruvic acid tris cyclohexylamine salt, 2-phosphonooxy-2-propenoic acid tri cyclohexylammonium salt, axi c 1/4 +/-ui feminineea ey>>. 1/4 masculine degrees .ni, phospho enol pyruvic acid tri cyclohexylammonium salt enzymatic | |
cyclohexanamine;2-phosphonooxyprop-2-enoic acid |
Specifications
35556-70-8 | |
C21H44N3O6P | |
1 g | |
MJKYGUXBFYGLLM-UHFFFAOYSA-N | |
cyclohexanamine;2-phosphonooxyprop-2-enoic acid | |
11754241 | |
≥98.0% (T) | |
Phosphoenolpyruvic Acid Tris(cyclohexylammonium) Salt Hydrate |
White | |
MFCD00004258 | |
phosphoenolpyruvic acid tris cyclohexylammonium salt, tris cyclohexylamine ; phosphoenolpyruvic acid, phosphoenolpyruvic acid, tris cyclohexylammonium salt, pep-3cha, phosphoenol pyruvate tri cyclohexylammonium salt, phosphoenolpyruvic acid tricyclohexylammonium salt, phosphoenolpyruvic acid tris cyclohexylamine salt, 2-phosphonooxy-2-propenoic acid tri cyclohexylammonium salt, axi c 1/4 +/-ui feminineea ey>>. 1/4 masculine degrees .ni, phospho enol pyruvic acid tri cyclohexylammonium salt enzymatic | |
C=C(C(=O)O)OP(=O)(O)O.C1CCC(CC1)N.C1CCC(CC1)N.C1CCC(CC1)N | |
465.572 | |
465.57 | |
Crystalline Powder |
Safety and Handling
TSCA : Yes
Recommended Storage : Refrigerator