accessibility menu, dialog, popup

UserName

NH2HOOH2COPOHOHONH2NH2

CAS RN 35556-70-8

IUPAC Name: 2-(phosphonooxy)prop-2-enoic acid; tris(cyclohexanamine)
Synonyms: tris(cyclohexylamine); phosphoenolpyruvic acid
Molecular Weight (g/mol): 465.57
Molecular Formula: C21H44N3O6P
InChi Key: MJKYGUXBFYGLLM-UHFFFAOYSA-N
SMILES: NC1CCCCC1.NC1CCCCC1.NC1CCCCC1.OC(=O)C(=C)OP(O)(O)=O
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results