missing translation for 'onlineSavingsMsg'
Learn More

Phosphoenolpyruvic Acid Tris(cyclohexylammonium) Salt Hydrate 98.0+%, TCI America™

Catalog No. P02561G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P02561G 1 g
P02565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P02561G Supplier TCI America Supplier No. P02561G

Chemical Identifiers

CAS 35556-70-8
Molecular Formula C21H44N3O6P
Molecular Weight (g/mol) 465.57
MDL Number MFCD00004258
InChI Key MJKYGUXBFYGLLM-UHFFFAOYSA-N
Synonym phosphoenolpyruvic acid tris cyclohexylammonium salt, tris cyclohexylamine ; phosphoenolpyruvic acid, phosphoenolpyruvic acid, tris cyclohexylammonium salt, pep-3cha, phosphoenol pyruvate tri cyclohexylammonium salt, phosphoenolpyruvic acid tricyclohexylammonium salt, phosphoenolpyruvic acid tris cyclohexylamine salt, 2-phosphonooxy-2-propenoic acid tri cyclohexylammonium salt, axi c 1/4 +/-ui feminineea ey>>. 1/4 masculine degrees .ni, phospho enol pyruvic acid tri cyclohexylammonium salt enzymatic
PubChem CID 11754241
IUPAC Name 2-(phosphonooxy)prop-2-enoic acid; tris(cyclohexanamine)
SMILES NC1CCCCC1.NC1CCCCC1.NC1CCCCC1.OC(=O)C(=C)OP(O)(O)=O

Specifications

Color White
Quantity 1 g
Formula Weight 465.57
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Phosphoenolpyruvic Acid Tris(cyclohexylammonium) Salt Hydrate
TSCA Yes
Recommended Storage Refrigerator