missing translation for 'onlineSavingsMsg'
Learn More

Phosphoenolpyruvic acid tricyclohexylammonium salt

Catalog No. AAJ6194314
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6194314 25 g
AAJ6194303 1 g
AAJ6194306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6194314 Supplier Thermo Scientific Chemicals Supplier No. J6194314

Metabolic intermediate in many kinase reactions | CAS: 35556-70-8 | C21H44N3O6P | 465.572 g/mol

Metabolic intermediate in many kinase reactions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 35556-70-8
Molecular Formula C21H44N3O6P
Molecular Weight (g/mol) 465.57
MDL Number MFCD00004258
InChI Key MJKYGUXBFYGLLM-UHFFFAOYSA-N
Synonym phosphoenolpyruvic acid tris cyclohexylammonium salt, tris cyclohexylamine ; phosphoenolpyruvic acid, phosphoenolpyruvic acid, tris cyclohexylammonium salt, pep-3cha, phosphoenol pyruvate tri cyclohexylammonium salt, phosphoenolpyruvic acid tricyclohexylammonium salt, phosphoenolpyruvic acid tris cyclohexylamine salt, 2-phosphonooxy-2-propenoic acid tri cyclohexylammonium salt, axi c 1/4 +/-ui feminineea ey>>. 1/4 masculine degrees .ni, phospho enol pyruvic acid tri cyclohexylammonium salt enzymatic
PubChem CID 11754241
IUPAC Name 2-(phosphonooxy)prop-2-enoic acid; tris(cyclohexanamine)
SMILES NC1CCCCC1.NC1CCCCC1.NC1CCCCC1.OC(=O)C(=C)OP(O)(O)=O

Specifications

Color White
Physical Form Powder
Quantity 25 g
Beilstein 3752336
Formula Weight 465.58
Percent Purity ≥99%
Chemical Name or Material Phosphoenolpyruvic acid tricyclohexylammonium salt
EINECSNumber 252-618-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only