missing translation for 'onlineSavingsMsg'
Learn More

Phosphoenolpyruvic acid monopotassium salt, 99%

CAS: 4265-07-0 | C3H4KO6P | 206.131 g/mol

Supplier:  Thermo Scientific Chemicals B2035814

 View more versions of this product

Catalog No. AAB2035814

Only null left
Explore available promotions

Description

Description

Phosphoenolpyruvic acid monopotassium salt is used in glycolysis and gluconeogeneis. It is used for regeneration of ATP from ADP in enzyme-catalyzed syntheses. It is metabolized by an enzyme namely pyruvate kinase to yield pyruvate. It acts as protection against cell injury induced by oxidative stress and against dysfunction in carbohydrate metabolism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phosphoenolpyruvic acid monopotassium salt is used in glycolysis and gluconeogeneis. It is used for regeneration of ATP from ADP in enzyme-catalyzed syntheses. It is metabolized by an enzyme namely pyruvate kinase to yield pyruvate. It acts as protection against cell injury induced by oxidative stress and against dysfunction in carbohydrate metabolism.

Solubility
Soluble in water. Slightly soluble in methanol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Specifications

Specifications

Phosphoenolpyruvic acid monopotassium salt
∼170°C (decomposition)
C3H4KO6P
25 g
Hygroscopic
Soluble in water. Slightly soluble in methanol.
C=C(C(=O)O)OP(=O)(O)[O-].[K+]
206.131
206.14
4265-07-0
0.99
MFCD00044476
4603446
potassium 1-carboxyvinyl hydrogenphosphate, potassium hydrogen 2-phosphonatooxy acrylate, 2-propenoic acid, 2-phosphonooxy-, monopotassium salt, potassium 1-carboxyvinyl hydrogen phosphate, phospho enol pyruvic acid monopotassium salt, phosphoenolpyruvic acid monopotassium salt, 2-propenoic acid, 2-phosphonooxy-, potassium salt 1:1, potassium 2-hydrogen phosphonooxy prop-2-enoic acid, phosphoenolpyruvate potassium salt, potassium 2-hydrogen phosphonatooxy prop-2-enoic acid
SOSDSEAIODNVPX-UHFFFAOYSA-M
potassium;1-carboxyethenyl hydrogen phosphate
23678879
99%

Chemical Identifiers

4265-07-0
206.131
SOSDSEAIODNVPX-UHFFFAOYSA-M
23678879
C=C(C(=O)O)OP(=O)(O)[O-].[K+]
C3H4KO6P
MFCD00044476
potassium 1-carboxyvinyl hydrogenphosphate, potassium hydrogen 2-phosphonatooxy acrylate, 2-propenoic acid, 2-phosphonooxy-, monopotassium salt, potassium 1-carboxyvinyl hydrogen phosphate, phospho enol pyruvic acid monopotassium salt, phosphoenolpyruvic acid monopotassium salt, 2-propenoic acid, 2-phosphonooxy-, potassium salt 1:1, potassium 2-hydrogen phosphonooxy prop-2-enoic acid, phosphoenolpyruvate potassium salt, potassium 2-hydrogen phosphonatooxy prop-2-enoic acid
potassium;1-carboxyethenyl hydrogen phosphate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 224-247-0

TSCA : Yes

Recommended Storage : Store at -20°C; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only