missing translation for 'onlineSavingsMsg'
Learn More

Phosphocholine Chloride Sodium Salt Hydrate 98.0+%, TCI America™

Catalog No. P08345G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P08345G 5 g
P08341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P08345G Supplier TCI America Supplier No. P08345G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16904-96-4
Molecular Formula C5H13ClNNa2O4P
Molecular Weight (g/mol) 263.57
MDL Number MFCD09037359
InChI Key HMCRHROSIPYRGL-UHFFFAOYSA-L
Synonym Phosphorylcholine Chloride Sodium Salt
PubChem CID 85635
IUPAC Name disodium chloride 2-(trimethylazaniumyl)ethyl phosphate
SMILES [Na+].[Na+].[Cl-].C[N+](C)(C)CCOP([O-])([O-])=O

Specifications

Color White
Quantity 5 g
Formula Weight 263.57
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Phosphocholine Chloride Sodium Salt Hydrate [for Cholinesterase Substrate]