accessibility menu, dialog, popup

UserName

CAS RN 16904-96-4

IUPAC Name: disodium chloride 2-(trimethylazaniumyl)ethyl phosphate
Synonyms: disodium chloride choline phosphate
Molecular Weight (g/mol): 263.57
Molecular Formula: C5H13ClNNa2O4P
InChi Key: HMCRHROSIPYRGL-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[Cl-].C[N+](C)(C)CCOP([O-])([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results