missing translation for 'onlineSavingsMsg'
Learn More

4-(trans-4-Propylcyclohexyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Numéro de catalogue. P21621G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
P21621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. P21621G Fournisseur TCI America Code fournisseur P21621G
Il en reste null

Identifiants chimiques

CAS 146862-02-4
Molecular Formula C15H23BO2
MDL Number MFCD06658197
InChI Key QTBUVZSHCNAPRT-UHFFFAOYSA-N
PubChem CID 3104710
IUPAC Name [4-(4-propylcyclohexyl)phenyl]boronic acid

Spécifications

Color White
Quantity 1 g
Synonym 4-trans-4-propylcyclohexyl phenylboronic acid, 4-trans-4-n-propylcyclohexyl phenyl boronic acid, 4-trans-4-propylcyclohexyl phenyl boronic acid, 4-4-propylcyclohexyl phenyl boronic acid, 4-4-propylcyclohexyl phenylboronic acid, boronic acid, 4-trans-4-propylcyclohexyl phenyl, 4-1s,4r-4-propylcyclohexyl phenylboronic acid, pubchem7891, 4-4-propylcyclohexyl phenylboronicacid
SMILES B(C1=CC=C(C=C1)C2CCC(CC2)CCC)(O)O
Molecular Weight (g/mol) 246.157
Formula Weight 246.16
Physical Form Crystalline Powder
Chemical Name or Material 4-(trans-4-Propylcyclohexyl)phenylboronic Acid (contains varying amounts of Anhydride)