accessibility menu, dialog, popup

UserName

CAS RN 146862-02-4

IUPAC Name: [4-(4-propylcyclohexyl)phenyl]boronic acid
Synonyms: 4-(4-propylcyclohexyl)phenylboronic acid
Molecular Weight (g/mol): 246.16
Molecular Formula: C15H23BO2
InChi Key: QTBUVZSHCNAPRT-UHFFFAOYSA-N
SMILES: CCCC1CCC(CC1)C1=CC=C(C=C1)B(O)O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results