Learn More
Phospho-Tau (Thr231) Recombinant Rabbit Monoclonal Antibody (1H6L6), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen LS701056
Description
Intact IgG appears on a non-reducing gel as ∽150 kDa band and upon reduction generating a ∽25 kDa light chain band and a ∽50 kDa heavy chain. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Phospho-Tau (Thr231) Recombinant Monoclonal antibody specifically detects Phospho-Tau (Thr231) in Human, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
| Phospho-Tau (Thr231) | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide | |
| AI413597; AW045860; DDPAC; FLJ31424; FTDP17; FTDP-17; G protein beta1/gamma2 subunit-interacting factor 1; map tau; Mapt; MAPTL; MGC138549; microtubule associated protein tau; microtubule-associated protein tau; microtubule-associated protein tau, isoform 4; microtubules; MSTD; Mtapt; MTBT1; MTBT2; Neurofibrillary tangle protein; neurofibrillary tangles; Neuronal Marker; paired helical filament-tau; PHFtau; PHF-tau; PPND; PPP1R103; protein phosphatase 1, regulatory subunit 103; pTau; RNPTAU; Tau; Tau microtubule-associated protein; tau protein; Tau-4; Tau5; Unknown (protein for MGC:134287) | |
| Phosphopeptide corresponding to amino acids 224â236 of human microtubule-associated protein tau | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 1H6L6 | |
| Unconjugated | |
| P10636, P19332 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 29477, 4137 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| IgG |