missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-Syndecan 4 (Ser179) Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5105801
Description
Antibody detects endogenous levels of Syndecan 4 only when phosphorylated at Ser179.
Syndecans are type I integral membrane proteoglycans that are involved in cell-extracellular matrix adhesion and growth factor binding. Syndecan 1 is a matrix receptor which binds to Collagens, Fibronectin and Thrombospondin. Syndecan 1 and Syndecan 3 interact with MK (midkine), a growth/differentiation factor involved in embryogenesis. Syndecan 2 is highly expressed at areas of high morphogenetic activity, such as epithelial-mesenchymal interfaces and the prechondrogenic and preosteogenic mesenchymal condensations. Syndecan 4 functions cooperatively with integrins in the processes of cell spreading and focal adhesion assembly.Chemical Identifiers
| Phospho-Syndecan 4 (Ser179) | |
| -20°C | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| Sdc4 | |
| AA959608; Amphiglycan; AW108331; MGC22217; RATRYUDOCA; RYUDOCA; ryudocan; ryudocan amphiglycan; ryudocan core protein; Ryudocan/syndecan 4; S4; SDC4; Synd4; syndecan 4; syndecan 4 (amphiglycan, ryudocan); syndecan proteoglycan 4; syndecan-4 | |
| Sdc4 | |
| Primary | |
| Sequential chromatography |
| 1 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O35988, P31431, P34901 | |
| 20971, 24771, 6385 | |
| A synthesized peptide derived from human SDC4(Accession P31431), corresponding to amino acid residues around phosphorylated Ser179. | |
| Antibody |
Specifications
| Phospho-Syndecan 4 (Ser179) | |
| Polyclonal | |
| Unconjugated | |
| Sdc4 | |
| AA959608; Amphiglycan; AW108331; MGC22217; RATRYUDOCA; RYUDOCA; ryudocan; ryudocan amphiglycan; ryudocan core protein; Ryudocan/syndecan 4; S4; SDC4; Synd4; syndecan 4; syndecan 4 (amphiglycan, ryudocan); syndecan proteoglycan 4; syndecan-4 | |
| Rabbit | |
| Sequential chromatography | |
| RUO | |
| 20971, 24771, 6385 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O35988, P31431, P34901 | |
| Sdc4 | |
| A synthesized peptide derived from human SDC4(Accession P31431), corresponding to amino acid residues around phosphorylated Ser179. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |