Learn More
Invitrogen™ Phospho-STING (Ser366) Recombinant Rabbit Monoclonal Antibody (PSH09-72)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA562891
Description
Positive Control: THP-1 treated with 80nM TPA for 16 hours then transfected with 5 μg/mL Poly (dA:dT) for 3 hours cell lysate.Tissue Specificity: Low tissue specificitySubcellular Location: Endoplasmic reticulum membrane, Cytoplasm, perinuclear region, Endoplasmic reticulum-Golgi intermediate compartment membrane, Golgi apparatus membrane, Cytoplasmic vesicle, autophagosome membrane, Mitochondrion outer membrane, Cell membrane.Sequence Similarities: 77% Mouse/RatPredicted band size: 42 kDa.
STING is a 379 amino acid containing signaling protein that acts as an important component of innate immune signaling. This signaling protein is able to activate both NF-kappa-B and IRF3 transcription pathways to induce expression of type I interferon (IFN-alpha and IFN-beta) and exert a potent anti-viral state following expression. STING may have translocon function, the translocon possibly being able to influence the induction of type I interferons and is also involved in transduction of apoptotic signals via its association with the major histocompatibility complex class II (MHC-II) thus facilitating the detection of intracellular viral RNA species as well as B-form DNA and mediates death signaling via activation of the extracellular signal-regulated kinase (ERK) pathway. Generally seen in endoplasmic reticulum membrane, cell membrane and mitochondrion outer membrane, it is ubiquitously expressed in most of tissues.Specifications
| Phospho-STING (Ser366) | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q86WV6 | |
| TMEM173 | |
| Synthetic phospho-peptide corresponding to residues surrounding Ser366 of Human STING. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Dot Blot, Immunoprecipitation, Western Blot | |
| PSH09-72 | |
| Unconjugated | |
| TMEM173 | |
| 2610307O08Rik; endoplasmic reticulum IFN stimulator; endoplasmic reticulum interferon stimulator; ERIS; hMITA; hSTING; hypothetical LOC533661; mediator of IRF3 activation; Mita; mitochondrial mediator of IRF3 activation; mitochondrial transmembrane protein 173; MMITA; Mpys; mSTING; NET23; N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner; poSTING; RGD1562552; rSTING; SAVI; Stimulator of interferon genes protein; stimulator of interferon response cGAMP interactor 1; STING; STING1; tm173; Tmem173; Transmembrane protein 173 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 340061 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |