missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-SMAD1/SMAD5 (Ser463, Ser465) Recombinant Rabbit Monoclonal Antibody (31H14L11)

Catalog No. 700047
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
700047 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 700047 Supplier Invitrogen™ Supplier No. LS700047
Only null left

Rabbit Recombinant Monoclonal Antibody

This antibody is predicted to react with mouse, primate, rat, Rhesus monkey , sea urchin, ovine, porcine, Xenopus and zebrafish based on sequence homology. Intact IgG appears on a non-reducing gel as ~150 kDa band and upon reduction generating a ~25 kDa light chain band and a ~50 kDa heavy chain. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

The Smad family of proteins are functioning in the transmission of extracellular signals in the TGF-beta signaling pathway. Binding of a TGF-beta superfamily ligands to extracellular receptors triggers phosphorylation of Smad2 at a Serine-Serine-Methionine-Serine (SSMS) motif at its C-terminus. Phosphorylated Smad2 is then able to form a complex with Smad4. These complexes accumulate in the cell nucleus, where they are directly participating in the regulation of gene expression. In mammals, eight Smad proteins have been identified to date. The Smad family of proteins can be divided into three functional groups: the receptor-activated Smads (R-Smads), common mediator Smads (Co-Smads), and the inhibitory Smads (I-Smads). The R-Smads are directly phosphorylated by the activated type I receptors on their C-terminal Ser-Ser-X-Ser (SSXS) motif and include Smad1, Smad2, Smad3, Smad5, and Smad8. Smad2 and Smad3 are phosphorylated in response to TGF-beta and activin, whereas Smad1, Smad5, and Smad8 are phosphorylated in response to BMP (Bone Morphogenetic Protein). This C-terminal phosphorylation allows R-Smad binding to Co-Smad, Smad4, and translocation to the nucleus where they regulate TGF-beta target genes. Smad6 and Smad7 belong to the I-Smad which bind to the type I receptor or Smad4 and block their interaction with R-Smads.

Specifications

Antigen Phospho-SMAD1/SMAD5 (Ser463, Ser465)
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 31H14L11
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene SMAD1
Gene Accession No. Q15797, Q99717
Gene Alias 1110051M15Rik; AI451355; AI528653; BSP1; BSP-1; dwarfin-A; Dwarfin-C; dwf-A; DWFC; Dwf-C; hSMAD1; hSmad5; JV41; JV4-1; JV5-1; MAD (mothers against decapentaplegic, Drosophila) homolog 1; MAD homolog 1; MAD homolog 5; MAD homolog1 (mothers against decapentaplegic, Drosophila); MAD, mothers against decapentaplegic homolog 1; MAD, mothers against decapentaplegic homolog 5; Madh1; Madh5; Madr1; mad-related protein 1; Mlp1; mMad1; mothers against decapentaplegic homolog 1; Mothers against decapentaplegic homolog 5; mothers against decapentaplegic, drosophila, homolog of, 5; mothers against DPP homolog 1; mothers against DPP homolog 5; mothers-against-DPP-related 1; Msmad5; MusMLP; OTTHUMP00000223331; SMA- and MAD-related protein 5; SMAD; Smad 1; SMAD 5; SMAD family member 1; SMAD family member 5; SMAD, mothers against DPP homolog 1; SMAD, mothers against DPP homolog 5; SMAD1; SMAD5; TGF-beta signaling protein 1; transforming growth factor-beta signaling protein 1; Transforming growth factor-beta-signaling protein 1
Gene Symbols SMAD1, Smad5
Host Species Rabbit
Immunogen A peptide corresponding to amino acids 456-465 of Q15797 and Q99717.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4086, 4090
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less