missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-Shc (Tyr427) Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104857
Description
Antibody detects endogenous levels of SHC only when phosphorylated at Tyrosine 427.
Skp2 (S-phase Kinase-associated Protein 2) belongs to the family of F-box proteins that interact with the Cyclin A-Cdk2 complex. Skp2 is essential for the G1-S transition in both transformed cells and diploid fibroblasts. Biochemical and genetic experiments have demonstrated that Skp2 is required for the ubiquitination and consequent degradation of p27 in cultured mammalian cells and in vitro reconstitution assays. In normal tissues, Skp2 is expressed in tonsil and placenta. In tumor tissues, Skp2 is expressed widely, in some colon, prostate, pancreas, and skin cancers, with high expression in lung, breast, ovarian, and endometrial cancers. Research suggests that Skp2 plays a role in oncogenesis, particularly in the pathogenesis of lymphomas and breast cancers.Chemical Identifiers
| Phospho-Shc (Tyr427) | |
| -20°C | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| SHC1 | |
| FLJ26504; OTTHUMP00000035472; p66; P66shc; RP11-307C12.1; SH2 domain protein C1; SHC; SHC (Src homology 2 domain containing) transforming protein 1; SHC (Src homology 2 domain-containing) transforming protein 1; SHC adaptor protein 1; SHC1; SHCA; SHC-transforming protein 1; SHC-transforming protein 3; SHC-transforming protein A; src homology 2 domain-containing transforming protein C1; src homology 2 domain-containing-transforming protein C1 | |
| SHC1 | |
| Primary | |
| sequential chromatography |
| 1 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P29353, P98083, Q5M824 | |
| 20416, 6464, 85385 | |
| A synthesized peptide derived from human SHC1(Accession P29353), corresponding to amino acid residues around phosphorylated Tyr427. | |
| Antibody |
Specifications
| Phospho-Shc (Tyr427) | |
| Polyclonal | |
| Unconjugated | |
| SHC1 | |
| FLJ26504; OTTHUMP00000035472; p66; P66shc; RP11-307C12.1; SH2 domain protein C1; SHC; SHC (Src homology 2 domain containing) transforming protein 1; SHC (Src homology 2 domain-containing) transforming protein 1; SHC adaptor protein 1; SHC1; SHCA; SHC-transforming protein 1; SHC-transforming protein 3; SHC-transforming protein A; src homology 2 domain-containing transforming protein C1; src homology 2 domain-containing-transforming protein C1 | |
| Rabbit | |
| sequential chromatography | |
| RUO | |
| 20416, 6464, 85385 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P29353, P98083, Q5M824 | |
| SHC1 | |
| A synthesized peptide derived from human SHC1(Accession P29353), corresponding to amino acid residues around phosphorylated Tyr427. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |