missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-SGLT1 (Ser335) Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5105325
Description
Antibody detects endogenous levels of SLC5A1 only when phosphorylated at Ser335.
Glucose transporters are integral membrane proteins that mediate the transport of glucose and structurally-related substances across cellular membranes. Two families of glucose transporter have been identified: the facilitated-diffusion glucose transporter family. The SLC5A1 gene encodes a protein that is involved in the active transport of glucose and galactose into eukaryotic and some prokaryotic cells.Chemical Identifiers
| Phospho-SGLT1 (Ser335) | |
| -20°C | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| Slc5a1 | |
| D22S675; High affinity sodium-glucose cotransporter; HSGLT1; MGC93553; Na; Na(+)/glucose cotransporter 1; Na+/glucose cotransporter 1; NAGT; Sglt1; SLC5A1; sodium glucose cotransporter 1; sodium/glucose cotransporter 1; sodium-glucose cotransporter 1; sodium-glucose cotransporter-like 1; solute carrier family 5 (sodium/glucose cotransporter), member 1; solute carrier family 5 member 1; solute carrier family 5, member 1; solute carrier family 5, member alpha 1; Solute carrier family 5, member alpha 1 (Na+/glucose cotransporter) | |
| Slc5a1 | |
| Primary | |
| Sequential Chromatography |
| 1 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P13866, P53790, Q8C3K6 | |
| 20537, 25552, 6523 | |
| A synthesized peptide derived from human SLC5A1(Accession P13866), corresponding to amino acid residues around phosphorylated Ser335. | |
| Antibody |
Specifications
| Phospho-SGLT1 (Ser335) | |
| Polyclonal | |
| Unconjugated | |
| Slc5a1 | |
| D22S675; High affinity sodium-glucose cotransporter; HSGLT1; MGC93553; Na; Na(+)/glucose cotransporter 1; Na+/glucose cotransporter 1; NAGT; Sglt1; SLC5A1; sodium glucose cotransporter 1; sodium/glucose cotransporter 1; sodium-glucose cotransporter 1; sodium-glucose cotransporter-like 1; solute carrier family 5 (sodium/glucose cotransporter), member 1; solute carrier family 5 member 1; solute carrier family 5, member 1; solute carrier family 5, member alpha 1; Solute carrier family 5, member alpha 1 (Na+/glucose cotransporter) | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 20537, 25552, 6523 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P13866, P53790, Q8C3K6 | |
| Slc5a1 | |
| A synthesized peptide derived from human SLC5A1(Accession P13866), corresponding to amino acid residues around phosphorylated Ser335. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |