missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-RyR2 (Ser2808) Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA536758

Catalog No. PIPA536758

Only null left
Explore available promotions

Description

Description

This antibody detects endogenous protein at a molecular weight of 565 kDa. Purity is >95% by SDS-PAGE.

Dihydropyridine receptor (DHPR) is a surface membrane protein critical for the excitation-contraction coupling of striated muscle. DHPR and the sarcoplasmic reticulum ryanodine receptor (RyR) are two key components of the intracellular junctions, where depolarization of the surface membrane is converted into the release of Ca2+ from internal stores. The alpha1-subunit of the DHPR contains a cytoplasmic loop which is thought to be involved in the interactions with RyR. Phosphorylation of the DHPR alpha1-subunit is also thought to play a role in the functional interaction of DHPR and RyR. Mutation in DHPR alpha1 results in excitation-contraction uncoupling, leading to muscular dysgenesis, a complete inactivity in developing skeletal muscles. Cells that do not express RyR also lack excitation-contraction coupling and exhibit a severalfold reduction in Ca2+ current density.
Specifications

Specifications

Phospho-RyR2 (Ser2808)
Polyclonal
Unconjugated
Ryr2
9330127I20Rik; ARVC2; ARVD2; calcium release channel; cardiac muscle ryanodine receptor; Cardiac muscle ryanodine receptor-calcium release channel; cardiac ryanodine receptor 2; cardiac-type ryanodine receptor; hRYR-2; islet-type ryanodine receptor; kidney-type ryanodine receptor; Ryanodine receptor; ryanodine receptor 2; ryanodine receptor 2 (cardiac); ryanodine receptor 2, cardiac; ryanodine receptor type 2; ryanodine receptor type II; RyR; RYR2; RYR-2; Type 2 ryanodine receptor; VTSIP
Rabbit
Antigen Affinity Chromatography
RUO
20191, 6262, 689560
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
B0LPN4, E9Q401, Q92736
Ryr2
Synthetic phosphopeptide derived from human RyR2 around the phosphorylation site of Serine 2808.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions