missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-RIP3 (Ser316) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105246
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105246 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105246 Supplier Invitrogen™ Supplier No. PA5105246
Only null left

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of RIPK3 only when phosphorylated at Ser316.

RIPK3 is a member of the receptor-interacting protein (RIP) family of serine/threonine protein kinases, which contains a C-terminal domain unique from other RIP family members. RIPK3 is predominantly localized to the cytoplasm, and specifically, are colocalized in the mitochondrion which can undergo nucleocytoplasmic shuttling dependent on novel nuclear localization and export signals. RIPK3 is a component of the tumor necrosis factor (TNF) receptor-I signaling complex, and can induce apoptosis and weakly activate the NF-kappaB transcription factor and caspase-8 prevents RIPK3-dependent necrosis without inducing apoptosis by functioning in a proteolytically active complex with CFLAR and that this complex is required for the protective function.

Specifications

Antigen Phospho-RIP3 (Ser316)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RIPK3
Gene Accession No. Q9Y572
Gene Alias 2610528K09Rik; AW107945; Hcyp2; Homocysteine respondent protein HCYP2; mRIP3; receptor interacting protein 3; receptor interacting serine/threonine kinase 3; receptor-interacting protein 3; receptor-interacting serine/threonine-protein kinase 3; receptor-interacting serine-threonine kinase 3; Rip3; RIP-3; Ripk3; RIP-like protein kinase 3
Gene Symbols RIPK3
Host Species Rabbit
Immunogen A synthesized peptide derived from human RIPK3(Accession Q9Y572), corresponding to amino acid residues around phosphorylated Ser316.
Purification Method Sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11035
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less