missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-p27 Kip1 (Thr157) Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA512650

Catalog No. PIPA512650


Only null left

Description

Description

Phospho-p27 Kip1 (Thr157) Polyclonal Antibody for IHC (P), DB

p27 Kip1 functions as a negative regulator of G1 mouse expression and has been proposed to function as a possible mediator of TGFb induced G1 arrest. p27 Kip1 is an important regulator of cell cycle progression, a potent inhibitor of cyclin E- and cyclin A-CDK2 complexes. p27 Kip1 forms a complex with cyclin type D-CDK4 complexes and is involved in the assembly, stability, and modulation of CCND1-CDK4 complex activation. p27 Kip1 acts either as an inhibitor or an activator of cyclin type D-CDK4 complexes depending on its phosphorylation state and/or stoichometry. p27 Kip1 has a molecular weight of approximately 27kDa and the degradation of this protein, which is triggered by its CDK dependent phosphorylation and subsequent ubiquitination by SCF complexes, is required for the cellular transition from quiescence to the proliferative state.
Specifications

Chemical Identifiers

Phospho-p27 Kip1 (Thr157)
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Polyclonal
Liquid
IgG
Human
Cdkn1b
AA408329; AI843786; Cdki1b; CDKN1B; CDKN4; CDN1B; cyclin dependent kinase inhibitor 1B; cyclin-dependent kinase inhibitor 1B; Cyclin-dependent kinase inhibitor 1B (p27 Kip1); Cyclin-dependent kinase inhibitor 1B (p27, Kip1); cyclin-dependent kinase inhibitor p27; Cyclin-dependent kinase inhibitor p27 Kip1; cyclin-dependent kinase inhibitor p27/Kip1; KIP1; MEN1B; MEN4; p27; P27 CDKN1B; p27(KIP1); p27Kip1
Cdkn1b
Primary
Antigen affinity chromatography, Protein A
0.4 mg/mL
Dot Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
RUO
PBS with 0.09% sodium azide; pH 7.4
P46527
1027
KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding T157 of human p27Kip1.
Antibody

Specifications

Phospho-p27 Kip1 (Thr157)
Polyclonal
Unconjugated
Cdkn1b
AA408329; AI843786; Cdki1b; CDKN1B; CDKN4; CDN1B; cyclin dependent kinase inhibitor 1B; cyclin-dependent kinase inhibitor 1B; Cyclin-dependent kinase inhibitor 1B (p27 Kip1); Cyclin-dependent kinase inhibitor 1B (p27, Kip1); cyclin-dependent kinase inhibitor p27; Cyclin-dependent kinase inhibitor p27 Kip1; cyclin-dependent kinase inhibitor p27/Kip1; KIP1; MEN1B; MEN4; p27; P27 CDKN1B; p27(KIP1); p27Kip1
Rabbit
Antigen affinity chromatography, Protein A
RUO
1027
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Dot Blot, Immunohistochemistry (Paraffin)
0.4 mg/mL
PBS with 0.09% sodium azide; pH 7.4
P46527
Cdkn1b
KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding T157 of human p27Kip1.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions