missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-p130Cas (Tyr410) Recombinant Rabbit Monoclonal Antibody (23GB3870)

Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA553656
Description
Phospho-p130Cas (Tyr410) Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot
The p130Cas (Cas for Crk-associated substrate) is a common cellular target of phosphorylation signal via v-Crk and v-Src oncoproteins. p130Cas has a unique structure that contains a Src homology (SH)-3 domain followed by multiple YXXP motifs and a proline-rich regionp130Cas is implicated in a variety of biological processes including cell adhesion, cell migration, growth factor stimulation, cytokine receptor engagement and bacterial infection. Physiological functions of p130Cas include cardiovascular development, actin filament assembly and Src-induced cell transformation.Chemical Identifiers
| Phospho-p130Cas (Tyr410) | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| BCAR1 | |
| AI385681; BCA1; Bcar1; BCAR1 scaffold protein, Cas family member; BCAR1, Cas family scaffold protein; BCAR1, Cas family scaffolding protein; breast cancer anti-estrogen resistance 1; breast cancer anti-estrogen resistance protein 1; Cas; Cas scaffolding protein family member 1; CAS1; CASS1; CRKAS; CRK-associated substrate; Crk-associated substrate p130Cas; OTTHUMP00000174941; p130 Cas; P130CAS; p130Cas (pTyr410); p130Cas (pY410); p130Cas phospho; phospho p130Cas; v-crk-associated tyrosine kinase substrate | |
| BCAR1 | |
| Primary | |
| Affinity chromatography |
| 23GB3870 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P56945, Q61140, Q63767 | |
| 12927, 25414, 9564 | |
| A synthetic peptide of human Phospho-BCAR1 (Tyr410) (403-417AA). | |
| Antibody |
Specifications
| Phospho-p130Cas (Tyr410) | |
| Recombinant Monoclonal | |
| Unconjugated | |
| BCAR1 | |
| AI385681; BCA1; Bcar1; BCAR1 scaffold protein, Cas family member; BCAR1, Cas family scaffold protein; BCAR1, Cas family scaffolding protein; breast cancer anti-estrogen resistance 1; breast cancer anti-estrogen resistance protein 1; Cas; Cas scaffolding protein family member 1; CAS1; CASS1; CRKAS; CRK-associated substrate; Crk-associated substrate p130Cas; OTTHUMP00000174941; p130 Cas; P130CAS; p130Cas (pTyr410); p130Cas (pY410); p130Cas phospho; phospho p130Cas; v-crk-associated tyrosine kinase substrate | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12927, 25414, 9564 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB3870 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P56945, Q61140, Q63767 | |
| BCAR1 | |
| A synthetic peptide of human Phospho-BCAR1 (Tyr410) (403-417AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |