missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-mTOR (Ser2448) Recombinant Mouse Monoclonal Antibody (MRRBY)

Mouse Recombinant Monoclonal Antibody

Supplier:  Invitrogen™ 740274M

 View more versions of this product

Catalog No. 740274M

Only null left

Description

Description

This is an engineered recombinant antibody that was developed to provide superior sensitivity without changing the paratope. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include better specificity, lot-to-lot consistency and improved signal-to-noise ratio. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

FRAP1 (mTOR) is a serine/threonine kinase that plays a critical role in cellular growth and proliferation. Perturbations in the mTOR/PI3-kinase/AKT pathway are associated with numerous forms of cancer. FRAP1 is also the target of rapamycin and its analogues, which are currently used as immunosuppressants and cancer therapeutics. Mutations affecting the gene results in Smith-Kingsmore syndrome.
Specifications

Chemical Identifiers

Phospho-mTOR (Ser2448)
1 mg/mL
Flow Cytometry, Western Blot
Unconjugated
Mouse
RUO
PBS with 0.09% sodium azide; pH 7.4
P42345
2475
Primary
Protein A/G
MRRBY
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Recombinant Monoclonal
Liquid
IgG2a κ
Human
MTOR
2610315D21Rik; AI327068; angiopoietin-like factor CDT6; FK506 binding protein 12-rapamycin associated protein 1; FK506 binding protein 12-rapamycin associated protein 2; FK506-binding protein 12-rapamycin complex-associated protein 1; FKBP12-rapamycin complex-associated protein; FKBP12-rapamycin complex-associated protein 1; FKBP-rapamycin associated protein; FKBP-rapamycin associated protein (FRAP); FKBP-rapamycin-associated protein FRAP; flat; FRAP; FRAP1; FRAP2; Mammalian target of rapamycin; mechanistic target of rapamycin; mechanistic target of rapamycin (serine/threonine kinase); mechanistic target of rapamycin kinase; Mtor; m-TOR; mTORC1; Raft1; Rapamycin and FKBP12 target 1; rapamycin and FKBP12 target-1 protein; rapamycin associated protein FRAP2; rapamycin target protein 1; RAPT1; serine/threonine-protein kinase mTOR; SKS
MTOR
Antibody

Specifications

Phospho-mTOR (Ser2448)
Recombinant Monoclonal
1 mg/mL
PBS with 0.09% sodium azide; pH 7.4
P42345
MTOR
Protein A/G
RUO
2475
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Flow Cytometry, Western Blot
MRRBY
Unconjugated
MTOR
2610315D21Rik; AI327068; angiopoietin-like factor CDT6; FK506 binding protein 12-rapamycin associated protein 1; FK506 binding protein 12-rapamycin associated protein 2; FK506-binding protein 12-rapamycin complex-associated protein 1; FKBP12-rapamycin complex-associated protein; FKBP12-rapamycin complex-associated protein 1; FKBP-rapamycin associated protein; FKBP-rapamycin associated protein (FRAP); FKBP-rapamycin-associated protein FRAP; flat; FRAP; FRAP1; FRAP2; Mammalian target of rapamycin; mechanistic target of rapamycin; mechanistic target of rapamycin (serine/threonine kinase); mechanistic target of rapamycin kinase; Mtor; m-TOR; mTORC1; Raft1; Rapamycin and FKBP12 target 1; rapamycin and FKBP12 target-1 protein; rapamycin associated protein FRAP2; rapamycin target protein 1; RAPT1; serine/threonine-protein kinase mTOR; SKS
Mouse
100 μL
Primary
Human
Antibody
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions