Learn More
Invitrogen™ Phospho-MAPKAPK2 (Thr334) Recombinant Rabbit Monoclonal Antibody (MAPKAPK2T334-H2), PE, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Supplier:  Invitrogen™ MA537287
 View more versions of this product
Catalog No. PIMA537287
Only null left
Description
Description
MAPKAPK2 mediates p38 and ERK signaling in vivo. It is known to be involved in many cellular processes including stress and inflammatory responses, nuclear export, gene expression regulation, and cell proliferation. MAPKAPK2 encodes a member of the Ser/Thr protein kinase family. MAPKAPK2 is regulated through direct phosphorylation by p38 MAP kinase. In conjunction with p38 MAP kinase, MAPKAPK2 is known to be involved in many cellular processes including stress and inflammatory responses, nuclear export, gene expression regulation and cell proliferation. Heat shock protein HSP27 was shown to be one of the substrates of this kinase in vivo. Two transcript variants encoding two different isoforms have been found for MAPKAPK2.
Specifications
Chemical Identifiers
Phospho-MAPKAPK2 (Thr334)
Flow Cytometry
PE
Rabbit
Human, Mouse
P49137, P49138
17164, 9261
A synthetic phospho-peptide corresponding to residues surrounding Thr334 of human phospho MAPKAPK-2.
Primary
4°C
MAPKAPK2
MAPKAPK2T334-H2
Recombinant Monoclonal
Liquid
RUO
PBS with 0.20% BSA and 0.09% sodium azide
AA960234; MAP kinase-activated protein kinase 2; MAPK activated protein kinase 2; MAPK-activated protein kinase 2; MAPKAP kinase 2; Mapkapk2; MAPKAP-K2; MAPKAPK-2; mitogen-activated protein kinase-activated protein kinase 2; MK2; MK-2; Rps6kc1
MAPKAPK2
IgG κ
Protein A/G
Antibody
Specifications
Phospho-MAPKAPK2 (Thr334)
Recombinant Monoclonal
PE
MAPKAPK2
AA960234; MAP kinase-activated protein kinase 2; MAPK activated protein kinase 2; MAPK-activated protein kinase 2; MAPKAP kinase 2; Mapkapk2; MAPKAP-K2; MAPKAPK-2; mitogen-activated protein kinase-activated protein kinase 2; MK2; MK-2; Rps6kc1
Rabbit
Protein A/G
RUO
17164, 9261
4°C
Liquid
Flow Cytometry
MAPKAPK2T334-H2
PBS with 0.20% BSA and 0.09% sodium azide
P49137, P49138
MAPKAPK2
A synthetic phospho-peptide corresponding to residues surrounding Thr334 of human phospho MAPKAPK-2.
10 Tests
Primary
Human, Mouse
Antibody
IgG κ
Product Suggestions
Product Suggestions