missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Multispecies JNK1/JNK2 (Phospho) [pT183/pY185] InstantOne™ ELISA Kit
Description
The InstantOne™ ELISA is specifically engineered for accurate measurement of phosphorylated human and mouse JNK 1/2 in cell lysates. Detection of phosphorylated rat JNK 1/2 is expected. The InstantOne™ ELISA kit allows for fast analysis of samples in approximately one hour. All reagents used in a traditional sandwich ELISA are added in solution to a plate followed by a wash step and detection with the TMB colorimetric substrate.
C-Jun N-terminal Kinase (JNK) is also known as Mitogen-activated protein kinase (MAPK8). It belongs to the MAPK superfamily of stress-activated protein kinases. MAPKs are Serine-threonine protein kinases that are activated in response to a variety of extracellular stimuli, and mediate signal transduction from the cell surface to the nucleus. JNK is involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. JNK pathways are activated by stress and inflammatory signals. JNK is expressed as ten different isoforms due to differential mRNA splicing. The predominant forms are JNK1 and JNK2.
Specifications
Specifications
| Accession Number | P45983,P45984,Q91Y86,Q9WTU6 |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Cell Lysates |
| Target Species | Mouse, Human |
| For Use With (Application) | ELISA |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 26419, 26420, 5599, 5601 |
| Gene Symbol | JNK-55,JNK2,JNK2A,JNK2ALPHA,JNK2B,JNK2BETA,PRKM9,SAPK,SAPK1a,p54a,p54aSAPK,JNK,JNK-46,JNK1,JNK1A2,JNK21B1/2,PRKM8,SAPK1,SAPK1c |
| Kit Contents | Capture Antibody, Detection Antibody Reagent, InstantOne™ Assay Plate, Cell Lysis Buffer, Enhancer Solution, Wash Buffer (10X), Substrate Reagent (TMB), Stop Solution, Positive Control Cell Lysate (lyophilized), Plate seal |
| Show More |