missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Jak2 (Tyr1007, Tyr1008) Recombinant Rabbit Monoclonal Antibody (JAK2Y10071008-PB6), PE, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
$351.64 - $864.98
Specifications
| Antigen | Phospho-Jak2 (Tyr1007, Tyr1008) |
|---|---|
| Clone | JAK2Y10071008-PB6 |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Conjugate | PE |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
PIMA537445
|
Invitrogen™
MA537445 |
10 Tests |
Each for $351.64
|
|
|||||
|
PIMA537197
|
Invitrogen™
MA537197 |
100 Tests |
Each for $864.98
|
|
|||||
Description
Janus kinase 2 (JAK2) is a protein tyrosine kinase involved in a specific subset of cytokine receptor signaling pathways and is mutated in polycythemia vera. This kinase contains the catalytic domain (JH1) of JAK2. The activation of JAKs can lead to the phosphorylation of STAT (signal transducers and activatiors of transcription) proteins, which dimerize and translocate to the nucleus. Once translocated to the nucleus, the STAT proteins can modify transcription of numerous genes, including interferon-stimulated genes. JAK2 is required for the IFN gamma-receptor complex initiation and JAK1 functions as an amplifier. Some studies have suggested that the role of JAK2 might be performed by Tyk2 and JAK3, if they were positioned correctly within the IFN gamma-receptor complex. Mutations affecting JAK2 can cause Budd-Chiari syndrome, Polycythemia vera, Thrombocythemia 3, myelofibrosis, or Acute myelogenous leukemia.Specifications
| Phospho-Jak2 (Tyr1007, Tyr1008) | |
| Flow Cytometry | |
| PE | |
| Rabbit | |
| Human | |
| O60674 | |
| 3717 | |
| A synthetic phospho-peptide corresponding to residues surrounding Tyr1007/Tyr1008 of human phospho Jak2. | |
| Primary | |
| 4°C | |
| JAK2 |
| JAK2Y10071008-PB6 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.20% BSA and 0.09% sodium azide | |
| EC 2.7.10.2; Fd17; JAK 2; JAK2; JAK-2; Janus kinase 2; Janus kinase 2 (a protein tyrosine kinase); JTK 10; JTK10; kinase Jak2; THCYT3; Tyrosine-protein kinase JAK2 | |
| JAK2 | |
| IgG κ | |
| Protein A/G | |
| Antibody |