missing translation for 'onlineSavingsMsg'
Learn More

Phospho-Jak2 (Tyr1007, Tyr1008) Recombinant Rabbit Monoclonal Antibody (JAK2Y10071008-PB6), APC, Invitrogen™

Rabbit Recombinant Monoclonal Antibody

$351.64 - $864.98

Specifications

Antigen Phospho-Jak2 (Tyr1007, Tyr1008)
Clone JAK2Y10071008-PB6
Applications Flow Cytometry
Classification Recombinant Monoclonal
Conjugate APC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PIMA537447
View Documents
Invitrogen™
MA537447
10 Tests
Each for $351.64
Only null left
 
PIMA537199
View Documents
Invitrogen™
MA537199
100 Tests
Each for $864.98
Only null left
 
Description

Description

Janus kinase 2 (JAK2) is a protein tyrosine kinase involved in a specific subset of cytokine receptor signaling pathways and is mutated in polycythemia vera. This kinase contains the catalytic domain (JH1) of JAK2. The activation of JAKs can lead to the phosphorylation of STAT (signal transducers and activatiors of transcription) proteins, which dimerize and translocate to the nucleus. Once translocated to the nucleus, the STAT proteins can modify transcription of numerous genes, including interferon-stimulated genes. JAK2 is required for the IFN gamma-receptor complex initiation and JAK1 functions as an amplifier. Some studies have suggested that the role of JAK2 might be performed by Tyk2 and JAK3, if they were positioned correctly within the IFN gamma-receptor complex. Mutations affecting JAK2 can cause Budd-Chiari syndrome, Polycythemia vera, Thrombocythemia 3, myelofibrosis, or Acute myelogenous leukemia.
Specifications

Specifications

Phospho-Jak2 (Tyr1007, Tyr1008)
Flow Cytometry
APC
Rabbit
Human
O60674
3717
A synthetic phospho-peptide corresponding to residues surrounding Tyr1007/Tyr1008 of human phospho Jak2.
Primary
4°C
JAK2
JAK2Y10071008-PB6
Recombinant Monoclonal
Liquid
RUO
PBS with 0.20% BSA and 0.09% sodium azide
EC 2.7.10.2; Fd17; JAK 2; JAK2; JAK-2; Janus kinase 2; Janus kinase 2 (a protein tyrosine kinase); JTK 10; JTK10; kinase Jak2; THCYT3; Tyrosine-protein kinase JAK2
JAK2
IgG κ
Protein A/G
Antibody
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'